Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(4-Boc-1-piperidinyl)-4-bromopyrazole, 98%

Click to view available options
Quantity:
250mg
1g
5g
877399-50-3
C13H20BrN3O2
330.226
MFCD09831980
IYNZAVDBHAQODX-UHFFFAOYSA-N
tert-butyl 4-4-bromo-1h-pyrazol-1-yl piperidine-1-carboxylate,1-boc-4-4-bromopyrazol-1-yl piperidine,4-4-bromopyrazol-1-yl piperidine-1-carboxylic acid tert-butyl ester,4-4-bromo-pyrazol-1-yl-piperidine-1-carboxylic acid tert-butyl ester,tert-butyl 4-4-bromopyrazol-1-yl piperidine-1-carboxylate,n-boc-4-4-bromopyrazol-1-yl piperidine,3-2-ethoxycarbonylacetyl piperidine-1-carboxylic acid tert-butyl ester,1-piperidinecarboxylicacid, 4-4-bromo-1h-pyrazol-1-yl-, 1,1-dimethylethyl ester,ksc658s5d,1-boc-4-4-bromo-1-pyrazolyl piperidine
45480278
tert-butyl 4-(4-bromopyrazol-1-yl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)N2C=C(C=N2)Br
877399-50-3
C13H20BrN3O2
330.226
MFCD09831980
IYNZAVDBHAQODX-UHFFFAOYSA-N
tert-butyl 4-4-bromo-1h-pyrazol-1-yl piperidine-1-carboxylate,1-boc-4-4-bromopyrazol-1-yl piperidine,4-4-bromopyrazol-1-yl piperidine-1-carboxylic acid tert-butyl ester,4-4-bromo-pyrazol-1-yl-piperidine-1-carboxylic acid tert-butyl ester,tert-butyl 4-4-bromopyrazol-1-yl piperidine-1-carboxylate,n-boc-4-4-bromopyrazol-1-yl piperidine,3-2-ethoxycarbonylacetyl piperidine-1-carboxylic acid tert-butyl ester,1-piperidinecarboxylicacid, 4-4-bromo-1h-pyrazol-1-yl-, 1,1-dimethylethyl ester,ksc658s5d,1-boc-4-4-bromo-1-pyrazolyl piperidine
45480278
tert-butyl 4-(4-bromopyrazol-1-yl)piperidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(CC1)N2C=C(C=N2)Br
Color White
Quantity 250mg
Beilstein 14511017
Formula Weight 330.23
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 1-(4-Boc-1-piperidinyl)-4-bromopyrazole

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning