Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-bromobutoxy)-4-nitrobenzene, 85%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
55502-03-9
C10H12BrNO3
274.11
MFCD00980317
DBRBCFJUEVSKGZ-UHFFFAOYSA-N
1-4-bromobutoxy-4-nitrobenzene,4-nitrophenoxybutyl bromide
2063669
1-(4-bromobutoxy)-4-nitrobenzene
C1=CC(=CC=C1[N+](=O)[O-])OCCCCBr
55502-03-9
C10H12BrNO3
274.11
MFCD00980317
DBRBCFJUEVSKGZ-UHFFFAOYSA-N
1-4-bromobutoxy-4-nitrobenzene,4-nitrophenoxybutyl bromide
2063669
1-(4-bromobutoxy)-4-nitrobenzene
C1=CC(=CC=C1[N+](=O)[O-])OCCCCBr
Melting Point 37°C to 40°C
Boiling Point 165°C to 170°C (0.5mmHg)
Color Beige to Brown
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 25g
CAS Min % 80.0
CAS Max % 100.0
Assay Percent Range 85%
Linear Formula NO2C6H4O(CH2)4Br
Formula Weight 274.11
Physical Form Adhering Chunks
Percent Purity ≥80%
Chemical Name or Material 1-(4-Bromobutoxy)-4-nitrobenzene
Show More Show Less

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.