Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4'-Bromophenoxy)-1-ethoxyethane (Tech.),90%, Acros Organics

Click to view available options
Quantity:
100g
39255-20-4
C10H13BrO2
245.116
MFCD00000096
QACZJJNHOATNHA-UHFFFAOYSA-N
1-bromo-4-2-ethoxyethoxy benzene,1-bromanyl-4-2-ethoxyethoxy benzene,benzene,1-bromo-4-2-ethoxyethoxy
21995844
1-bromo-4-(2-ethoxyethoxy)benzene
CCOCCOC1=CC=C(C=C1)Br
39255-20-4
C10H13BrO2
245.116
MFCD00000096
QACZJJNHOATNHA-UHFFFAOYSA-N
1-bromo-4-2-ethoxyethoxy benzene,1-bromanyl-4-2-ethoxyethoxy benzene,benzene,1-bromo-4-2-ethoxyethoxy
21995844
1-bromo-4-(2-ethoxyethoxy)benzene
CCOCCOC1=CC=C(C=C1)Br
Infrared Spectrum Authentic
Color Green-Yellow
Quantity 100g
Refractive Index 1.5210 to 1.5230 (20°C, 589nm)
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Linear Formula BrC6H4OCH(CH3)OCH2CH3
Formula Weight 245.12
Physical Form Liquid
Percent Purity ≥88%
Chemical Name or Material 1-(4'-Bromophenoxy)-1-ethoxyethane, tech., 90%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.