Alfa Aesar™ 1-(4-Bromophenyl)-2,5-dimethylpyrrole

Manufacturer: Alfa Aesar™ H5059406

Quantity 5g

 View more versions of this product

Catalog No. AAH5059406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 5044-24-6
    Molecular Formula C12H12BrN
    Formula Weight 250.14
    MDL Number MFCD00277042
    Chemical Name or Material 1-(4-Bromophenyl)-2,5-dimethylpyrrole

    Specifications

    Formula Weight 250.14
    Molecular Formula C12H12BrN
    MDL Number MFCD00277042
    CAS 5044-24-6
    Quantity 5g
    Chemical Name or Material 1-(4-Bromophenyl)-2,5-dimethylpyrrole

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit