Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(4-Bromophenyl)ethanol, 95%

Click to view available options
Quantity:
5g
5391-88-8
C8H9BrO
201.063
MFCD00004514
XTDTYSBVMBQIBT-UHFFFAOYSA-N
1-4-bromophenyl ethanol,4-bromo-alpha-methylbenzyl alcohol,1-4-bromophenyl ethan-1-ol,4-bromophenyl methyl carbinol,p-bromophenylmethylcarbinol,4-bromophenylmethylcarbinol,4-bromo-a-methylbenzyl alcohol,benzenemethanol, 4-bromo-alpha-methyl,benzyl alcohol, p-bromo-.alpha.-methyl,benzenemethanol, 4-bromo-.alpha.-methyl
95352
1-(4-bromophenyl)ethanol
CC(C1=CC=C(C=C1)Br)O
5391-88-8
C8H9BrO
201.063
MFCD00004514
XTDTYSBVMBQIBT-UHFFFAOYSA-N
1-4-bromophenyl ethanol,4-bromo-alpha-methylbenzyl alcohol,1-4-bromophenyl ethan-1-ol,4-bromophenyl methyl carbinol,p-bromophenylmethylcarbinol,4-bromophenylmethylcarbinol,4-bromo-a-methylbenzyl alcohol,benzenemethanol, 4-bromo-alpha-methyl,benzyl alcohol, p-bromo-.alpha.-methyl,benzenemethanol, 4-bromo-.alpha.-methyl
95352
1-(4-bromophenyl)ethanol
CC(C1=CC=C(C=C1)Br)O
Boiling Point 119°C to 121°C (7mmHg)
Flash Point 63°C (145°F)
Melting Point 36°C to 38°C
Quantity 5g
Beilstein 2042029
Formula Weight 201.06
Percent Purity 95%
Chemical Name or Material 1-(4-Bromophenyl)ethanol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning