Alfa Aesar™ 1-(4-Bromophenyl)pyrrolidin-2-one, 98%

Manufacturer: Alfa Aesar™ H5056306

Quantity 5g

 View more versions of this product

Catalog No. AAH5056306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7661-32-7
    Molecular Formula C10H10BrNO
    Formula Weight 240.1
    MDL Number MFCD00138502
    Chemical Name or Material 1-(4-Bromophenyl)pyrrolidin-2-one

    Specifications

    Formula Weight 240.1
    Boiling Point 98° to 101°C
    Molecular Formula C10H10BrNO
    MDL Number MFCD00138502
    CAS 7661-32-7
    Quantity 5g
    Chemical Name or Material 1-(4-Bromophenyl)pyrrolidin-2-one
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit