Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Butanediol, 99%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
110-63-4
C4H10O2
90.12
WERYXYBDKMZEQL-UHFFFAOYSA-N
butane-1,4-diol
OCCCCO
110-63-4
C4H10O2
90.12
WERYXYBDKMZEQL-UHFFFAOYSA-N
butane-1,4-diol
OCCCCO

B2144, 110-63-4, C4H10O2

Spectrum™ 1,4-Butanediol or tetramethylene glycol is a colorless viscous liquid. The majority of this material is used in the production of tetrahydrofuran for conversion into Spandex. However, there are illicit uses of the material and the CA DOJ has put this material on the 21-day precursor hold list for California customers. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Amber Glass Bottle
Quantity 4 L
CAS Max % 1
Percent Purity 99%