Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Butanediphosphonic acid, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass Bottle
4671-77-6
C4H12O6P2
218.082
MFCD01631212
JKTORXLUQLQJCM-UHFFFAOYSA-N
1,4-butanediphosphonic acid,1,4-butylenediphosphonic acid,1,4-diphosphonobutane,tetramethylene phosphonic acid,butane-1,4-bisphosphonic acid,tetramethylenebisphosphonic acid,butane-1,4-diyldiphosphonic acid
437082
4-phosphonobutylphosphonic acid
C(CCP(=O)(O)O)CP(=O)(O)O
4671-77-6
C4H12O6P2
218.082
MFCD01631212
JKTORXLUQLQJCM-UHFFFAOYSA-N
1,4-butanediphosphonic acid,1,4-butylenediphosphonic acid,1,4-diphosphonobutane,tetramethylene phosphonic acid,butane-1,4-bisphosphonic acid,tetramethylenebisphosphonic acid,butane-1,4-diyldiphosphonic acid
437082
4-phosphonobutylphosphonic acid
C(CCP(=O)(O)O)CP(=O)(O)O
Packaging Glass Bottle
Melting Point 219.0°C to 221.0°C
Color White
Quantity 1g
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (O)P(OH)2(CH2)4P(OH)2(O)
Formula Weight 218.08
Physical Form Fine Powder
Percent Purity 98%
Chemical Name or Material 1, 4-Butanediphosphonic acid
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA