Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(4-Chloro-2-nitrophenyl)pyrrolidine, 97%

Click to view available options
Quantity:
250mg
1g
5g
41173-36-8
C10H11ClN2O2
226.66
QATZMIJXHKOELL-UHFFFAOYSA-N
1-4-chloro-2-nitrophenyl pyrrolidine,1-4-chloro-2-nitro-phenyl pyrrolidine,n-4-chloro-2-nitrophenyl pyrrolidine,4-chloro-2-nitrophenyl pyrrolidine,qatzmijxhkoell-uhfffaoysa,1-4-chloro-2-nitro-phenyl-pyrrolidine,1-4-chloro-2-nitrophenyl pyrrolidine #,1-4-chloranyl-2-nitro-phenyl pyrrolidine,2-pyrrolidin-1-yl-5-chloro-1-nitrobenzene
285321
1-(4-chloro-2-nitrophenyl)pyrrolidine
C1CCN(C1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
41173-36-8
C10H11ClN2O2
226.66
QATZMIJXHKOELL-UHFFFAOYSA-N
1-4-chloro-2-nitrophenyl pyrrolidine,1-4-chloro-2-nitro-phenyl pyrrolidine,n-4-chloro-2-nitrophenyl pyrrolidine,4-chloro-2-nitrophenyl pyrrolidine,qatzmijxhkoell-uhfffaoysa,1-4-chloro-2-nitro-phenyl-pyrrolidine,1-4-chloro-2-nitrophenyl pyrrolidine #,1-4-chloranyl-2-nitro-phenyl pyrrolidine,2-pyrrolidin-1-yl-5-chloro-1-nitrobenzene
285321
1-(4-chloro-2-nitrophenyl)pyrrolidine
C1CCN(C1)C2=C(C=C(C=C2)Cl)[N+](=O)[O-]
Quantity 5g
Beilstein 915044
Formula Weight 226.66
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 1-(4-Chloro-2-nitrophenyl)pyrrolidine

TSCA : No

Recommended Storage : Ambient temperatures