Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-methanol, 98%

Click to view available options
Quantity:
250mg
1g
5g
133902-66-6
C9H8ClN3O
209.633
MFCD00140928
CAHIFLPAMJOAGI-UHFFFAOYSA-N
1-4-chlorophenyl-1h-1,2,3-triazol-4-yl methanol,1-4-chlorophenyl-1h-1,2,3-triazol-4-yl methanol,1-4-chlorophenyl-1h-1,2,3-triazol-4-yl methanol,1-4-chlorophenyl triazol-4-yl methanol,1-4-chlorophenyl-1,2,3-triazol-4-yl methanol,1-4-chlorophenyl-1h-1,2,3-triazole-4-yl-methanol,1-4-chlorophenyl-1,2,3-triazol-4-yl methan-1-ol,pubchem13003,chlorophenyltriazolylmethanol,1-4-chlorophenyl-1h-1,2,3-triazol-4-ylmethanol
2763878
[1-(4-chlorophenyl)triazol-4-yl]methanol
C1=CC(=CC=C1N2C=C(N=N2)CO)Cl
133902-66-6
C9H8ClN3O
209.633
MFCD00140928
CAHIFLPAMJOAGI-UHFFFAOYSA-N
1-4-chlorophenyl-1h-1,2,3-triazol-4-yl methanol,1-4-chlorophenyl-1h-1,2,3-triazol-4-yl methanol,1-4-chlorophenyl-1h-1,2,3-triazol-4-yl methanol,1-4-chlorophenyl triazol-4-yl methanol,1-4-chlorophenyl-1,2,3-triazol-4-yl methanol,1-4-chlorophenyl-1h-1,2,3-triazole-4-yl-methanol,1-4-chlorophenyl-1,2,3-triazol-4-yl methan-1-ol,pubchem13003,chlorophenyltriazolylmethanol,1-4-chlorophenyl-1h-1,2,3-triazol-4-ylmethanol
2763878
[1-(4-chlorophenyl)triazol-4-yl]methanol
C1=CC(=CC=C1N2C=C(N=N2)CO)Cl
Quantity 250mg
Melting Point 141°C to 145°C
Beilstein 4250389
Formula Weight 209.63
Physical Form Solid
Percent Purity 98%
Chemical Name or Material 1-(4-Chlorophenyl)-1H-1,2,3-triazole-4-methanol