Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea, 97%, Alfa Aesar™

Click to view available options
Quantity:
250mg
1g
369-77-7
C14H9Cl2F3N2O
349.134
MFCD00867294
ZFSXZJXLKAJIGS-UHFFFAOYSA-N
cloflucarban,halocarban,irgasan cf3,cloflucarbon,trifluoromethyldichlorocarbanilide,unii-i5zzy3dc5g,cloflucarban usan,4,4'-dichloro-3-trifluoromethyl carbanilide,i5zzy3dc5g,halocarban inn
9719
1-(4-chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
C1=CC(=CC=C1NC(=O)NC2=CC(=C(C=C2)Cl)C(F)(F)F)Cl
369-77-7
C14H9Cl2F3N2O
349.134
MFCD00867294
ZFSXZJXLKAJIGS-UHFFFAOYSA-N
cloflucarban,halocarban,irgasan cf3,cloflucarbon,trifluoromethyldichlorocarbanilide,unii-i5zzy3dc5g,cloflucarban usan,4,4'-dichloro-3-trifluoromethyl carbanilide,i5zzy3dc5g,halocarban inn
9719
1-(4-chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
C1=CC(=CC=C1NC(=O)NC2=CC(=C(C=C2)Cl)C(F)(F)F)Cl
Quantity 1g
Formula Weight 349.13
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 1-(4-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea