Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(4-Chlorophenyl)piperazine 98%, Acros Organics

Click to view available options
Quantity:
5g
38212-33-8
C10H13ClN2
196.678
UNEIHNMKASENIG-UHFFFAOYSA-N
1-4-chlorophenyl piperazine,piperazine, 1-4-chlorophenyl,1-4-chloro-phenyl-piperazine,para-chlorophenylpiperazine,pcpp,1-4-chlorophenyl-piperazine,1-4-chlorophenyl piperazine,p-chlorophenylpiperazine,4-chlorophenyl piperazine,1-4-chlorphenyl piperazin
97478
1-(4-chlorophenyl)piperazine
C1CN(CCN1)C2=CC=C(C=C2)Cl
38212-33-8
C10H13ClN2
196.678
UNEIHNMKASENIG-UHFFFAOYSA-N
1-4-chlorophenyl piperazine,piperazine, 1-4-chlorophenyl,1-4-chloro-phenyl-piperazine,para-chlorophenylpiperazine,pcpp,1-4-chlorophenyl-piperazine,1-4-chlorophenyl piperazine,p-chlorophenylpiperazine,4-chlorophenyl piperazine,1-4-chlorphenyl piperazin
97478
1-(4-chlorophenyl)piperazine
C1CN(CCN1)C2=CC=C(C=C2)Cl
Quantity 5g
Color Yellow
Melting Point 70°C to 77°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Formula Weight 196.68
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 1-(4-Chlorophenyl)piperazine, 98%