Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Cineole, 85%, Acros Organics™

Click to view available options
Quantity:
25g
100g
500g
470-67-7
C10H18O
154.25
RFFOTVCVTJUTAD-UHFFFAOYSA-N
1,4-cineole,isocineole,1,4-cineol,1,4-epoxy-p-menthane,isocineple,1,4-cineole natural,p-menthane, 1,4-epoxy,unii-b55jtu839b,fema no. 3658
10106
CHEBI:80788
1-methyl-4-propan-2-yl-7-oxabicyclo[2.2.1]heptane
CC(C)C12CCC(O1)(CC2)C
470-67-7
C10H18O
154.25
RFFOTVCVTJUTAD-UHFFFAOYSA-N
1,4-cineole,isocineole,1,4-cineol,1,4-epoxy-p-menthane,isocineple,1,4-cineole natural,p-menthane, 1,4-epoxy,unii-b55jtu839b,fema no. 3658
10106
CHEBI:80788
1-methyl-4-propan-2-yl-7-oxabicyclo[2.2.1]heptane
CC(C)C12CCC(O1)(CC2)C
Flash Point 47°C
Boiling Point 65°C (21.0 hPa)
Density 0.8870g/mL
Infrared Spectrum Authentic
Specific Gravity 0.887
Melting Point -46.0°C
Quantity 500g
Assay Percent Range 80% min. (GC)
Formula Weight 154.25
Percent Purity 85%
Chemical Name or Material 1,4-Cineole
Show More Show Less

GHS H Statement
Flammable liquid and vapor.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Store in a well-ventilated place.
Keep container tightly closed.

GHS Signal Word: Warning

EINECSNumber : 207-428-9