Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,4-Cyclohexanedimethanol, cis + trans, 99%

Click to view available options
Quantity:
100g
500g
105-08-8
C8H16O2
144.214
MFCD00001512
YIMQCDZDWXUDCA-UHFFFAOYSA-N
1,4-cyclohexanedimethanol,trans-1,4-cyclohexanedimethanol,chdm,1,4-bis hydroxymethyl cyclohexane,rikabinol dm,cyclohexane-1,4-dimethanol,1,4-chidm,1,4-dimethylolcyclohexane,4-hydroxymethyl cyclohexyl methanol,1,4-cyclohexanedimethanol, cis
7735
[4-(hydroxymethyl)cyclohexyl]methanol
C1CC(CCC1CO)CO
105-08-8
C8H16O2
144.214
MFCD00001512
YIMQCDZDWXUDCA-UHFFFAOYSA-N
1,4-cyclohexanedimethanol,trans-1,4-cyclohexanedimethanol,chdm,1,4-bis hydroxymethyl cyclohexane,rikabinol dm,cyclohexane-1,4-dimethanol,1,4-chidm,1,4-dimethylolcyclohexane,4-hydroxymethyl cyclohexyl methanol,1,4-cyclohexanedimethanol, cis
7735
[4-(hydroxymethyl)cyclohexyl]methanol
C1CC(CCC1CO)CO
Applications
CHDM is a precursor to polyesters. It is one of the most important co-monomers for production of polyethylene terephthalate (PET), or polyethylene terephthalic ester (PETE).

Solubility
Miscible with water, glycolether and ketones.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.
Density 1.04
Boiling Point 282°C to 284°C
Flash Point 161°C (322°F)
Quantity 100g
Melting Point ~30°C
Beilstein 1902271
Solubility Information Miscible with water,glycolether and ketones.
Formula Weight 144.21
Percent Purity 99%
Chemical Name or Material 1,4-Cyclohexanedimethanol, cis + trans

P264b-P280i-P305+P351+P338

H319

EINECSNumber : 203-268-9

RTECSNumber : GU9800000

TSCA : Yes

Recommended Storage : Ambient temperatures