Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Cyclohexanedione mono(2,2-dimethyltrimethylene ketal), 98+%, Acros Organics™

Click to view available options
Quantity:
10g
69225-59-8
C11H18O3
198.26
COKVDTKAWIFNTH-UHFFFAOYSA-N
3,3-dimethyl-1,5-dioxaspiro 5.5 undecan-9-one,1,4-cyclohexanedione mono-2,2-dimethyltrimethylene ketal,1,4-cyclohexanedione mono 2,2-dimethyltrimethylene ketal,3,3-dimethyl-1,5-dioxa-spiro 5.5 undecan-9-one,9,9-dimethyl-7,11-dioxaspiro 5.5 undecan-3-one,1,5-dioxaspiro 5.5 undecan-9-one, 3,3-dimethyl,pubchem11226,maybridge1_004236,1,4-cyclohexanedionemono-2,2-dimethyltrimethyleneketal
587968
3,3-dimethyl-1,5-dioxaspiro[5.5]undecan-9-one
CC1(COC2(CCC(=O)CC2)OC1)C
69225-59-8
C11H18O3
198.26
COKVDTKAWIFNTH-UHFFFAOYSA-N
3,3-dimethyl-1,5-dioxaspiro 5.5 undecan-9-one,1,4-cyclohexanedione mono-2,2-dimethyltrimethylene ketal,1,4-cyclohexanedione mono 2,2-dimethyltrimethylene ketal,3,3-dimethyl-1,5-dioxa-spiro 5.5 undecan-9-one,9,9-dimethyl-7,11-dioxaspiro 5.5 undecan-3-one,1,5-dioxaspiro 5.5 undecan-9-one, 3,3-dimethyl,pubchem11226,maybridge1_004236,1,4-cyclohexanedionemono-2,2-dimethyltrimethyleneketal
587968
3,3-dimethyl-1,5-dioxaspiro[5.5]undecan-9-one
CC1(COC2(CCC(=O)CC2)OC1)C
Quantity 10g
Flash Point >110°C
Melting Point 49.0°C to 50.0°C
Assay Percent Range 98% min. (GC)
Formula Weight 198.26
Percent Purity 98+%
Chemical Name or Material 1, 4-Cyclohexanedione mono(2, 2-dimethyltrimethylene ketal)

GHS Signal Word: Warning

EINECSNumber : 273-918-4