Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,4-Diacetoxy-2-butyne, 97%

Click to view available options
Quantity:
5g
25g
1573-17-7
C8H10O4
170.164
MFCD00014982
TVIMIQVBDDNCCR-UHFFFAOYSA-N
1,4-diacetoxy-2-butyne,but-2-yne-1,4-diyl diacetate,butynediol diacetate,2-butyne-1,4-diol diacetate,2-butyne-1,4-diol, diacetate,2-butyne-1,4-diol, 1,4-diacetate,4-acetyloxy but-2-yn-1-yl acetate,acmc-209dfx,2-butyne-1, diacetate,bis acetoxymethyl acetylene
74081
4-acetyloxybut-2-ynyl acetate
CC(=O)OCC#CCOC(=O)C
1573-17-7
C8H10O4
170.164
MFCD00014982
TVIMIQVBDDNCCR-UHFFFAOYSA-N
1,4-diacetoxy-2-butyne,but-2-yne-1,4-diyl diacetate,butynediol diacetate,2-butyne-1,4-diol diacetate,2-butyne-1,4-diol, diacetate,2-butyne-1,4-diol, 1,4-diacetate,4-acetyloxy but-2-yn-1-yl acetate,acmc-209dfx,2-butyne-1, diacetate,bis acetoxymethyl acetylene
74081
4-acetyloxybut-2-ynyl acetate
CC(=O)OCC#CCOC(=O)C
Boiling Point 116°C to 118°C (8mmHg)
Density 1.125
Refractive Index 1.454
Melting Point 27°C to 28°C
Flash Point >110°C (230°F)
Quantity 5g
Linear Formula CH3CO2CH2C≡CCH2O2CCH3
Formula Weight 170.16
Percent Purity 97%
Chemical Name or Material 1,4-Diacetoxy-2-butyne

EINECSNumber : 216-394-4

TSCA : Yes

Recommended Storage : Ambient temperatures