Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diaminoanthraquinone 90.0+%, TCI America™

Click to view available options
Quantity:
25g
500g
128-95-0
C14H10N2O2
238.246
MFCD00001224
FBMQNRKSAWNXBT-UHFFFAOYSA-N
1,4-diaminoanthraquinone,disperse violet 1,krisolamine,9,10-anthracenedione, 1,4-diamino,grasol violet r,seacyl violet r,acetate red violet r,1,4-diamino anthraquinone,setacyl violet r,oil violet r
31420
1,4-diaminoanthracene-9,10-dione
C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)N)N
128-95-0
C14H10N2O2
238.246
MFCD00001224
FBMQNRKSAWNXBT-UHFFFAOYSA-N
1,4-diaminoanthraquinone,disperse violet 1,krisolamine,9,10-anthracenedione, 1,4-diamino,grasol violet r,seacyl violet r,acetate red violet r,1,4-diamino anthraquinone,setacyl violet r,oil violet r
31420
1,4-diaminoanthracene-9,10-dione
C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)N)N
Melting Point 270°C
Color Black-Purple
Quantity 500g
Formula Weight 238.25
Physical Form Crystal-Powder at 20°C
Percent Purity ≥90.0% (HPLC,N)
Chemical Name or Material 1,4-Diaminoanthraquinone

EINECSNumber : (4)-0710&(5)-2540

RTECSNumber : CB6300000

TSCA : Yes