Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Diaminobutane dihydrochloride, 99+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
333-93-7
C4H12N2·2HCl
161.08
MFCD00012526
XXWCODXIQWIHQN-UHFFFAOYSA-N
1,4-diaminobutane dihydrochloride,putrescine dihydrochloride,tetramethylenediamine dihydrochloride,putrescine hydrochloride,1,4-butanediamine dihydrochloride,unii-x45sur7rhy,1,4-butanediamine, dihydrochloride,1,4-butanediamine, hydrochloride,1,4-diaminobutane, dihydrochloride,butane-1,4-diamine dihydrochloride
9532
butane-1,4-diamine;dihydrochloride
C(CCN)CN.Cl.Cl
333-93-7
C4H12N2·2HCl
161.08
MFCD00012526
XXWCODXIQWIHQN-UHFFFAOYSA-N
1,4-diaminobutane dihydrochloride,putrescine dihydrochloride,tetramethylenediamine dihydrochloride,putrescine hydrochloride,1,4-butanediamine dihydrochloride,unii-x45sur7rhy,1,4-butanediamine, dihydrochloride,1,4-butanediamine, hydrochloride,1,4-diaminobutane, dihydrochloride,butane-1,4-diamine dihydrochloride
9532
butane-1,4-diamine;dihydrochloride
C(CCN)CN.Cl.Cl
Packaging Glass bottle
Melting Point 280.0°C
Quantity 5g
Infrared Spectrum Authentic
Color White
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula H2N(CH2)4NH2·2HCl
Beilstein 04, 264
Merck Index 15, 8058
Solubility Information Solubility in water: soluble.
Formula Weight 161.08
Physical Form Crystalline Powder
Percent Purity 99+%
Chemical Name or Material 1, 4-Diaminobutane dihydrochloride
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF IN EYES: Rinse cautious

GHS Signal Word: Warning

EINECSNumber : 206-375-9