Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,4-Diaminobutane dihydrochloride, 99+%, ACROS Organics™
Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
Specifications
Infrared Spectrum | Authentic |
Quantity | 25g |
Packaging | Glass bottle |
Melting Point | 280.0°C |
Color | White |
CAS Min % | 99.0 |
CAS Max % | 100.0 |
Assay Percent Range | 99+% |
Linear Formula | H2N(CH2)4NH2·2HCl |
Beilstein | 04, 264 |
Show More |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
IF IN EYES: Rinse cautious
GHS Signal Word: Warning
EINECSNumber : 206-375-9