Alfa Aesar™ 1,4-Dibenzoylbenzene, 99%

Manufacturer: Alfa Aesar™ A1207014

Quantity 25g

 View more versions of this product

Catalog No. AAA1207014

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3016-97-5
    Molecular Formula C20H14O2
    Formula Weight 286.33
    MDL Number MFCD00014087
    Beilstein 1880440
    Synonym 4-Benzoylbenzophenone
    Chemical Name or Material 1,4-Dibenzoylbenzene

    Specifications

    Formula Weight 286.33
    Molecular Formula C20H14O2
    MDL Number MFCD00014087
    CAS 3016-97-5
    Melting Point 161° to 165°C
    Quantity 25g
    Beilstein 1880440
    Synonym 4-Benzoylbenzophenone
    Chemical Name or Material 1,4-Dibenzoylbenzene
    Assay 99%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit