Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,4-Dibenzyloxybenzene, 98%

Click to view available options
Quantity:
10g
50g
250g
621-91-0
C20H18O2
290.362
MFCD00016875
DYULYMCXVSRUPB-UHFFFAOYSA-N
1,4-dibenzyloxybenzene,1,4-bis benzyloxy benzene,hydroquinone dibenzyl ether,benzene, 1,4-bis phenylmethoxy,1,4-bis phenylmethoxy benzene,p-dibenzyloxybenzene,hydrochinon-dibenzylather,1,4-di benzyloxy benzene,acmc-209n0i,ksc493o6b
69315
1,4-bis(phenylmethoxy)benzene
C1=CC=C(C=C1)COC2=CC=C(C=C2)OCC3=CC=CC=C3
621-91-0
C20H18O2
290.362
MFCD00016875
DYULYMCXVSRUPB-UHFFFAOYSA-N
1,4-dibenzyloxybenzene,1,4-bis benzyloxy benzene,hydroquinone dibenzyl ether,benzene, 1,4-bis phenylmethoxy,1,4-bis phenylmethoxy benzene,p-dibenzyloxybenzene,hydrochinon-dibenzylather,1,4-di benzyloxy benzene,acmc-209n0i,ksc493o6b
69315
1,4-bis(phenylmethoxy)benzene
C1=CC=C(C=C1)COC2=CC=C(C=C2)OCC3=CC=CC=C3
Melting Point 125°C to 128°C
Odor Odorless
Quantity 50g
Beilstein 2058196
Formula Weight 290.36
Percent Purity 98%
Chemical Name or Material 1,4-Dibenzyloxybenzene