Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 1,4-Dibenzyloxybenzene, 98%
Click to view available options
Quantity:
10g
50g
250g
Specifications
Odor | Odorless |
Quantity | 250g |
Melting Point | 125°C to 128°C |
Beilstein | 2058196 |
Formula Weight | 290.36 |
Percent Purity | 98% |
Chemical Name or Material | 1,4-Dibenzyloxybenzene |
Safety and Handling
EINECSNumber : 210-714-6
TSCA : No
Recommended Storage : Ambient temperatures