Alfa Aesar™ 1,4-Dibromo-2,3-butanedione, 98%

Manufacturer: Alfa Aesar™ B2517014

Quantity 25g

 View more versions of this product

Catalog No. AAB2517014

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 6305-43-7
    Molecular Formula C4H4Br2O2
    Formula Weight 243.89
    MDL Number MFCD00000205
    UN Number UN3261
    Beilstein 970154
    Synonym alpha,alpha'-Dibromobiacetyl; 1,4-Dibromodiacetyl
    Chemical Name or Material 1,4-Dibromo-2,3-butanedione

    Specifications

    Formula Weight 243.89
    TSCA TSCA
    Melting Point 116° to 119°C
    Quantity 25g
    Molecular Formula C4H4Br2O2
    Odor Irritating
    MDL Number MFCD00000205
    CAS 6305-43-7
    UN Number UN3261
    Beilstein 970154
    Synonym alpha,alpha'-Dibromobiacetyl; 1,4-Dibromodiacetyl
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    RTECS Number EK2850000
    Chemical Name or Material 1,4-Dibromo-2,3-butanedione
    Assay 98%

    Description

    Applications
    1,4-Dibromo-2,3-butanedione is used in refluxing methanol yielded a new chiral pyridylthiazole ligand. It is also used as pharmaceutical intermediate.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents and bases.

    GHS H Statement
    H301-H314-H318
    Toxic if swallowed.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit