Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

Alfa Aesar™ 1,4-Dibromo-2,3-butanedione, 98%

Manufacturer:  Alfa Aesar™ B2517022

Quantity 100g

 View more versions of this product

Catalog No. AAB2517022

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Description

Applications
1,4-Dibromo-2,3-butanedione is used in refluxing methanol yielded a new chiral pyridylthiazole ligand. It is also used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents and bases.
Description & Specifications

Chemical Identifiers

6305-43-7
243.89
UN3261
alpha,alpha'-Dibromobiacetyl; 1,4-Dibromodiacetyl
C4H4Br2O2
MFCD00000205
970154
1,4-Dibromo-2,3-butanedione

Specifications

6305-43-7
100g
Irritating
MFCD00000205
970154
1,4-Dibromo-2,3-butanedione
116° to 119°C
C4H4Br2O2
243.89
UN3261
alpha,alpha'-Dibromobiacetyl; 1,4-Dibromodiacetyl
98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : EK2850000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit