Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,4-Dibromo-2-ethylbenzene, 96%

Click to view available options
Quantity:
1g
5g
289039-53-8
C8H8Br2
263.96
MFCD00672998
OMHJVVBCCMLFMY-UHFFFAOYSA-N
2,5-dibromoethylbenzene,acmc-20ap2i,1,4-dibromo-2-ethyl-benzene
18947383
1,4-dibromo-2-ethylbenzene
CCC1=C(C=CC(=C1)Br)Br
289039-53-8
C8H8Br2
263.96
MFCD00672998
OMHJVVBCCMLFMY-UHFFFAOYSA-N
2,5-dibromoethylbenzene,acmc-20ap2i,1,4-dibromo-2-ethyl-benzene
18947383
1,4-dibromo-2-ethylbenzene
CCC1=C(C=CC(=C1)Br)Br
Quantity 5g
Formula Weight 263.96
Percent Purity 96%
Chemical Name or Material 1,4-Dibromo-2-ethylbenzene

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning