Alfa Aesar™ 1,4-Dibromobutane, 98+%

Manufacturer: Alfa Aesar™ A1166909

Quantity 10g

 View more versions of this product

Catalog No. AAA1166909

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 110-52-1
    Molecular Formula C4H8Br2
    Linear Formula Br(CH2)4Br
    Formula Weight 215.93
    MDL Number MFCD00000261
    Beilstein 1071199
    Synonym Tetramethylene dibromide
    Chemical Name or Material 1,4-Dibromobutane

    Specifications

    Formula Weight 215.93
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    TSCA TSCA
    Health Hazard 1 Warning
    RTECS Number EJ7565000
    Boiling Point 194° to 196°C
    Molecular Formula C4H8Br2
    MDL Number MFCD00000261
    CAS 110-52-1
    Beilstein 1071199
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Flash Point 131°C (267°F)
    Melting Point −20°C
    Quantity 10g
    Linear Formula Br(CH2)4Br
    Synonym Tetramethylene dibromide
    Chemical Name or Material 1,4-Dibromobutane
    Assay ≥98%

    Description

    Applications
    1,4-Dibromobutane is used as an intermediate involved in the synthesis of active pharmaceutical ingredient and other organic compounds. It is also used in the investigation of metabolism of two halopropanes such as 1,3-dichloropropane and 2,2-dichloropropane. Further, it acts as a reagent to prepare diazadioxa oxovanadium(IV) macrocyclic complexes.

    Solubility
    Immiscible with water.

    Notes
    Incompatible with strong oxidizing agents and strong bases.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit