Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,4-Dichloro-2-nitrobenzene, 98%

Manufacturer:  Alfa Aesar™ A1189622

Quantity 100g

 View more versions of this product

Catalog No. AAA1189622


  Check Availability
Add to cart

Specifications

Description

Applications
1,4-Dichloro-2-nitrobenzene is a reagent used in the synthesis of antitrypanosomal, antileishmanial and antimalarial agents. It is also used in the synthesis of lysophosphatidic acid acyltransferase-β in hibitors.

Solubility
Soluble in water, ethanol, ether, benzene, carbon disulfide. Slightly soluble in carbon tetrachloride.

Notes
Incompatible with strong oxidizing agents, strong bases.
Description & Specifications

Chemical Identifiers

89-61-2
192
UN3077
2,5-Dichloronitrobenzene; Nitro-p-dichlorobenzene
C6H3Cl2NO2
MFCD00007074
778109
1,4-Dichloro-2-nitrobenzene

Specifications

100g
C6H3Cl2NO2
MFCD00007074
266° to 269°C
53° to 57°C
2,5-Dichloronitrobenzene; Nitro-p-dichlorobenzene
98%
89-61-2
192
UN3077
778109
125°C (257°F)
1,4-Dichloro-2-nitrobenzene

Safety and Handling

SDS & Safety Documents

GHS H Statement
H341-H302-H319
Suspected of causing genetic defects.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
P280h-P273-P305+P351+P338
Wear protective gloves/clothing.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : CZ5260000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit