Alfa Aesar™ 1,4-Dichloro-2-nitrobenzene, 98%

Manufacturer: Alfa Aesar™ A1189630

Quantity 250g

 View more versions of this product

Catalog No. AAA1189630

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 89-61-2
    Molecular Formula C6H3Cl2NO2
    Formula Weight 192
    MDL Number MFCD00007074
    UN Number UN3077
    Beilstein 778109
    Synonym 2,5-Dichloronitrobenzene; Nitro-p-dichlorobenzene
    Chemical Name or Material 1,4-Dichloro-2-nitrobenzene

    Specifications

    Formula Weight 192
    Health Hazard 3 GHS P Statement
    P280h-P273-P305+P351+P338
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    CAS 89-61-2
    Health Hazard 1 Warning
    RTECS Number CZ5260000
    Molecular Formula C6H3Cl2NO2
    UN Number UN3077
    Beilstein 778109
    Flash Point 125°C (257°F)
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Quantity 250g
    TSCA TSCA
    Melting Point 53° to 57°C
    Synonym 2,5-Dichloronitrobenzene; Nitro-p-dichlorobenzene
    MDL Number MFCD00007074
    Boiling Point 266° to 269°C
    Health Hazard 2 GHS H Statement
    H341-H302-H319
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Causes serious eye irritation.
    Chemical Name or Material 1,4-Dichloro-2-nitrobenzene
    Assay 98%

    Description

    Applications
    1,4-Dichloro-2-nitrobenzene is a reagent used in the synthesis of antitrypanosomal, antileishmanial and antimalarial agents. It is also used in the synthesis of lysophosphatidic acid acyltransferase-β in hibitors.

    Solubility
    Soluble in water, ethanol, ether, benzene, carbon disulfide. Slightly soluble in carbon tetrachloride.

    Notes
    Incompatible with strong oxidizing agents, strong bases.

    GHS H Statement
    H341-H302-H319
    Suspected of causing genetic defects.
    Harmful if swallowed.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P273-P305+P351+P338
    Wear protective gloves/clothing.
    Avoid release to the environment.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit