Alfa Aesar™ 1,4-Dichlorophthalazine, 98%

Manufacturer: Alfa Aesar™ H3329503

Quantity 1g

 View more versions of this product

Catalog No. AAH3329503

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 4752-10-7
    Molecular Formula C8H4Cl2N2
    Formula Weight 199.04
    MDL Number MFCD00006909
    Chemical Name or Material 1,4-Dichlorophthalazine

    Specifications

    Formula Weight 199.04
    Molecular Formula C8H4Cl2N2
    MDL Number MFCD00006909
    Recommended Storage −30° to −10°C
    Melting Point 164° to 166°C
    Quantity 1g
    CAS 4752-10-7
    Chemical Name or Material 1,4-Dichlorophthalazine
    Assay 98%

    Description

    Applications
    It is used as pharmaceutical intermediate. Benzoylthiophenes are allosteric enhancers (AE) of agonist activity at the A1 adenosine receptor.

    Solubility
    Sparingly soluble in water.(0.26 g/L) (25°C),

    Notes
    Store in cool dry place. Ensure proper ventilation. Incompatible with oxidizing agents.

    −30° to −10°C

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit