Learn More
1,4-Diethylbenzene, 98%, Thermo Scientific Chemicals
Description
1,4-Diethylbenzene, is used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics andcross-linking agents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,4-Diethylbenzene, is used as a solvent and an intermediate in the manufacture of other organic compounds for the finished products of such as curing agents, engineering plastics andcross-linking agents.
Solubility
It is insoluble in water. But soluble in ethanol, benzene,carbon tetrachloride.
Notes
Stable under ordinary conditions. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | -43°C |
Density | 0.862 |
Boiling Point | 182°C to 184°C |
Flash Point | 55°C (131°F) |
Odor | Aromatic |
Refractive Index | 1.495 |
Quantity | 5 g |
UN Number | UN2049 |
Beilstein | 1903396 |
Solubility Information | It is insoluble in water. But soluble in ethanol,benzene,carbon tetrachloride. |
Show More |
Safety and Handling
Hazard Category | H226-H304-H315-H318 |
Hazard Statement | GHS H Statement H226 Flammable liquid and vapour. |
Precautionary Statement | P210-P233-P240-P241-P242-P243-P264b-P280-P301+P310-P302+P352-P305+P351+P338-P310-P331-P332+P313-P362-P374-P380-P501c |
DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DIETHYLBENZENE |
EINECSNumber | 203-265-2 |
RTECSNumber | CZ5640700 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.