Alfa Aesar™ 1,4-Difluoro-2-iodobenzene, 97%

Manufacturer: Alfa Aesar™ H2689003

Quantity 1g

 View more versions of this product

Catalog No. AAH2689003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2265-92-1
    Molecular Formula C6H3F2I
    Formula Weight 239.99
    MDL Number MFCD03094151
    Chemical Name or Material 1,4-Difluoro-2-iodobenzene

    Specifications

    Formula Weight 239.99
    Health Hazard 3 GHS P Statement
    P210-P280-P305+P351+P338-P362-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 178° to 179°C
    Molecular Formula C6H3F2I
    MDL Number MFCD03094151
    CAS 2265-92-1
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H227
    Causes skin irritation.
    Causes serious eye irritation.
    Combustible liquid.
    Flash Point 85°C (185°F)
    Quantity 1g
    Chemical Name or Material 1,4-Difluoro-2-iodobenzene
    Assay 97%

    GHS H Statement
    H315-H319-H227
    Causes skin irritation.
    Causes serious eye irritation.
    Combustible liquid.

    GHS P Statement
    P210-P280-P305+P351+P338-P362-P403+P235-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Take off contaminated clothing and wash before reuse.
    Store in a well-ventilated place. Keep cool.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit