Alfa Aesar™ 1,4-Difluoro-2-nitrobenzene, 98%

Manufacturer: Alfa Aesar™ A1180714

Quantity 25g

 View more versions of this product

Catalog No. AAA1180714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 364-74-9
    Molecular Formula C6H3F2NO2
    Formula Weight 159.09
    MDL Number MFCD00007054
    UN Number UN2810
    Beilstein 2210200
    Synonym 2,5-Difluoronitrobenzene
    Chemical Name or Material 1,4-Difluoro-2-nitrobenzene

    Specifications

    Formula Weight 159.09
    Health Hazard 2 GHS H Statement
    H331-H373-H227-H302-H312-H315-H319-H335
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Combustible liquid.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P260-P261-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    RTECS Number CZ5715000
    MDL Number MFCD00007054
    Beilstein 2210200
    Health Hazard 1 Danger
    Melting Point −12°C
    Molecular Formula C6H3F2NO2
    CAS 364-74-9
    UN Number UN2810
    Synonym 2,5-Difluoronitrobenzene
    Flash Point 90°C (194°F)
    Boiling Point 206° to 207°C
    Quantity 25g
    Chemical Name or Material 1,4-Difluoro-2-nitrobenzene
    Assay 98%

    GHS H Statement
    H331-H373-H227-H302-H312-H315-H319-H335
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Combustible liquid.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P260-P261-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit