Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,4-Difluorobenzene, 99%

Click to view available options
Quantity:
25g
100g
540-36-3
C6H4F2
114.095
MFCD00000344
QUGUFLJIAFISSW-UHFFFAOYSA-N
p-difluorobenzene,benzene, 1,4-difluoro,para-difluorobenzene,benzene, p-difluoro,2,5-difluorobenzene,unii-y9fq2ef0z6,1,4-difluorbenzol,y9fq2ef0z6,benzene,4-difluoro,pubchem2309
10892
CHEBI:38585
1,4-difluorobenzene
C1=CC(=CC=C1F)F
540-36-3
C6H4F2
114.095
MFCD00000344
QUGUFLJIAFISSW-UHFFFAOYSA-N
p-difluorobenzene,benzene, 1,4-difluoro,para-difluorobenzene,benzene, p-difluoro,2,5-difluorobenzene,unii-y9fq2ef0z6,1,4-difluorbenzol,y9fq2ef0z6,benzene,4-difluoro,pubchem2309
10892
CHEBI:38585
1,4-difluorobenzene
C1=CC(=CC=C1F)F
Density 1.166
Refractive Index 1.441
Melting Point -13°C
Flash Point 2°C (35°F)
Odor Characteristic
Quantity 100g
Boiling Point 88°C to 89°C
UN Number UN1993
Beilstein 1904541
Merck Index 14,3148
Formula Weight 114.09
Percent Purity 99%
Chemical Name or Material 1,4-Difluorobenzene
Show More Show Less

GHS H Statement
H225-H319
Highly flammable liquid and vapour.
Causes serious eye irritation.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H225

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 208-742-9

RTECSNumber : CZ5658000

TSCA : No

Recommended Storage : Ambient temperatures