Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,4-Difluorobenzene, 99%

Manufacturer:  Alfa Aesar™ A1621714

Quantity 25g

 View more versions of this product

Catalog No. AAA1621714


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

540-36-3
114.09
UN1993
14,3148
1,4-Difluorobenzene
C6H4F2
MFCD00000344
1904541
p-Difluorobenzene

Specifications

2°C (35°F)
88° to 89°C
25g
C6H4F2
MFCD00000344
1904541
p-Difluorobenzene
99%
Characteristic
−13°C
540-36-3
114.09
UN1993
14,3148
1,4-Difluorobenzene

Safety and Handling

GHS H Statement
H225-H319
Highly flammable liquid and vapour.
Causes serious eye irritation.

GHS P Statement
P210-P260-P233-P243-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
Keep container tightly closed.
Take precautionary measures against static discharge.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

RTECSNumber : CZ5658000

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit