Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,4-Dihydroxyanthraquinone, 95%

Manufacturer:  Alfa Aesar™ A110100I

Quantity 5000g

 View more versions of this product

Catalog No. AAA110100I


  Check Availability
Add to cart

Specifications

Description

Applications
1,4-Dihydroxyanthraquinone is used to color gasoline and some heating oils. It is used as an intermediate for the synthesis of indanthrene- and alizarin-derived dyes. It is also used to form lake pigments with calcium, barium and lead. It may be used in the synthesis of cyclopentanoids by cyclization of α,β-unsaturated aldehyde with 1, 4 1,4-Dihydroxyanthraquinone. It has been studied as a long range emissive ratiometric fluorescent probe for live cell imaging.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Description & Specifications

Chemical Identifiers

81-64-1
240.21
1914036
C.I. 58050; Quinizarin
C14H8O4
MFCD00001209
14,8064
1,4-Dihydroxyanthraquinone

Specifications

222°C (431°F)
195° to 198°C
81-64-1
240.21
1914036
C.I. 58050; Quinizarin
95%
≈450°C
5000g
C14H8O4
MFCD00001209
14,8064
1,4-Dihydroxyanthraquinone

Safety and Handling

SDS & Safety Documents

RTECSNumber : CB6600000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit