Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dihydroxyanthraquinone 96%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
1kg
Packaging:
Glass bottle
81-64-1
C14H8O4
240.21
MFCD00001209
GUEIZVNYDFNHJU-UHFFFAOYSA-N
quinizarin,1,4-dihydroxyanthraquinone,quinizarine,chinizarin,solvent orange 86,1,4-dihydroxyanthrachinon,smoke orange r,1,4-dihydroxy-9,10-anthraquinone,macrolex orange gg,9,10-anthracenedione, 1,4-dihydroxy
6688
CHEBI:37487
1,4-dihydroxyanthracene-9,10-dione
C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)O
81-64-1
C14H8O4
240.21
MFCD00001209
GUEIZVNYDFNHJU-UHFFFAOYSA-N
quinizarin,1,4-dihydroxyanthraquinone,quinizarine,chinizarin,solvent orange 86,1,4-dihydroxyanthrachinon,smoke orange r,1,4-dihydroxy-9,10-anthraquinone,macrolex orange gg,9,10-anthracenedione, 1,4-dihydroxy
6688
CHEBI:37487
1,4-dihydroxyanthracene-9,10-dione
C1=CC=C2C(=C1)C(=O)C3=C(C=CC(=C3C2=O)O)O
Infrared Spectrum Authentic
Packaging Glass bottle
Color Brown-Red to Orange
Melting Point 191.0°C to 193.0°C
Boiling Point 450.0°C
Flash Point 222°C
Quantity 100g
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Beilstein 08, 450
Merck Index 15, 8180
Solubility Information Solubility in water: <1g/L (20°C). Other solubilities: soluble in hot water,soluble in alcohol,ether,benzene,sulfuric,acid,potassium chloride,chloroform,acetone,,aqueous alkali and other organic solvents
Formula Weight 240.21
Physical Form Crystalline Powder
Percent Purity 96%
Chemical Name or Material 1, 4-Dihydroxyanthraquinone, 96%
Show More Show Less

EINECSNumber : 201-368-7

RTECSNumber : CB6600000

TSCA : TSCA