Learn More
1,4-Dihydroxynaphthalene, 1:1 mixture with 1,4-Naphthoquinone, ACROS Organics™
Specifications
Color | Brown to Gray |
Infrared Spectrum | Authentic |
Quantity | 25g |
Linear Formula | C10H6(OH)2 |
Beilstein | 06, 979 |
Formula Weight | 160.17 |
Physical Form | Powder |
Chemical Name or Material | 1, 4-Dihydroxynaphthalene |
Safety and Handling
GHS H Statement:
Toxic in contact with skin.
Fatal if inhaled.
May cause respiratory irritation.
May cause an allergic skin reaction.
Causes skin irritation.
Toxic if swallowed.
Causes serious eye damage.
Very toxic to aquatic life.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger