Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,4-Diiodooctafluorobutane, 97%, stab. with copper

Click to view available options
Quantity:
5g
25g
375-50-8
C4F8I2
453.84
MFCD00042263
JILAKKYYZPDQBE-UHFFFAOYSA-N
1,4-diiodooctafluorobutane,octafluoro-1,4-diiodobutane,1,4-diiodoperfluorobutane,perfluoro-1,4-diiodobutane,1,4-diiodo octafluorobutane,butane, 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodo,acmc-1aigi,i cf2 4i,1,1,2,2,3,3,4,4-octafluoro-1,4-diiodo-butane
78993
1,1,2,2,3,3,4,4-octafluoro-1,4-diiodobutane
C(C(C(F)(F)I)(F)F)(C(F)(F)I)(F)F
375-50-8
C4F8I2
453.84
MFCD00042263
JILAKKYYZPDQBE-UHFFFAOYSA-N
1,4-diiodooctafluorobutane,octafluoro-1,4-diiodobutane,1,4-diiodoperfluorobutane,perfluoro-1,4-diiodobutane,1,4-diiodo octafluorobutane,butane, 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodo,acmc-1aigi,i cf2 4i,1,1,2,2,3,3,4,4-octafluoro-1,4-diiodo-butane
78993
1,1,2,2,3,3,4,4-octafluoro-1,4-diiodobutane
C(C(C(F)(F)I)(F)F)(C(F)(F)I)(F)F
Boiling Point 85°C (100mmHg)
Melting Point -9°C
Quantity 25g
Linear Formula I(CF2)4I
Beilstein 1777548
Formula Weight 453.84
Percent Purity 97%
Chemical Name or Material 1,4-Diiodooctafluorobutane, stabilized with copper

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning