Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,4-Diisopropylcyclohexane, cis + trans, 98%

Click to view available options
Quantity:
5g
25g
22907-72-8
C12H24
168.324
MFCD00019366
GMUJGUXGVJZHSK-UHFFFAOYSA-N
1,4-diisopropylcyclohexane,1,4-diisopropyl cyclohexane,1,4-di-isopropylcyclohexane,1,4-di propan-2-yl cyclohexane,cyclohexane,1,4-bis 1-methylethyl-,trans,acmc-1cbxy,1,4-diisopr opylcyclohexane,1,4-bis methylethyl cyclohexane,1alpha,4beta-diisopropylcyclohexane,1,4-diisopropylcyclohexane,cis+trans
140972
1,4-di(propan-2-yl)cyclohexane
CC(C)C1CCC(CC1)C(C)C
22907-72-8
C12H24
168.324
MFCD00019366
GMUJGUXGVJZHSK-UHFFFAOYSA-N
1,4-diisopropylcyclohexane,1,4-diisopropyl cyclohexane,1,4-di-isopropylcyclohexane,1,4-di propan-2-yl cyclohexane,cyclohexane,1,4-bis 1-methylethyl-,trans,acmc-1cbxy,1,4-diisopr opylcyclohexane,1,4-bis methylethyl cyclohexane,1alpha,4beta-diisopropylcyclohexane,1,4-diisopropylcyclohexane,cis+trans
140972
1,4-di(propan-2-yl)cyclohexane
CC(C)C1CCC(CC1)C(C)C
Boiling Point 214°C to 216°C
Quantity 5g
UN Number UN1993
Formula Weight 168.32
Percent Purity 98%
Chemical Name or Material 1,4-Diisopropylcyclohexane, cis + trans

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.