Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dimethoxybenzene, 99+%, ACROS Organics™

Click to view available options
Quantity:
100g
500g
1kg
Packaging:
Glass bottle
150-78-7
C8H10O2
138.17
MFCD00008401
OHBQPCCCRFSCAX-UHFFFAOYSA-N
p-dimethoxybenzene,p-methoxyanisole,hydroquinone dimethyl ether,benzene, 1,4-dimethoxy,quinol dimethyl ether,benzene, p-dimethoxy,dimethylhydroquinone ether,dimethyl ether hydroquinone,1,4-dimethoxybenzol,usaf an-9
9016
1,4-dimethoxybenzene
COC1=CC=C(C=C1)OC
150-78-7
C8H10O2
138.17
MFCD00008401
OHBQPCCCRFSCAX-UHFFFAOYSA-N
p-dimethoxybenzene,p-methoxyanisole,hydroquinone dimethyl ether,benzene, 1,4-dimethoxy,quinol dimethyl ether,benzene, p-dimethoxy,dimethylhydroquinone ether,dimethyl ether hydroquinone,1,4-dimethoxybenzol,usaf an-9
9016
1,4-dimethoxybenzene
COC1=CC=C(C=C1)OC
Boiling Point 213°C
Flash Point 98°C
Specific Gravity 1.05
Viscosity 1.04 mPa.s (65°C)
Melting Point 54°C to 58°C
Infrared Spectrum Authentic
Quantity 1kg
Packaging Glass bottle
Color White
Density 1.0500g/mL
CAS Min % 99.0
CAS Max % 100.0
Assay Percent Range 99+%
Linear Formula C6H4(OCH3)2
Beilstein 06, 843
Solubility Information Solubility in water: 0.8g/L (20 c). Other solubilities: soluble in benzene,alcohol,ethylacetate and,acetone,1g/10mL dioxane
Formula Weight 138.17
Physical Form Crystalline Powder, Crystals, or Flakes
Percent Purity ≥99.0%
Chemical Name or Material 1, 4-Dimethoxybenzene, 99+%
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 205-771-9

RTECSNumber : CZ6650000

TSCA : TSCA