Alfa Aesar™ 1,4-Dimethylpiperazine, 98%

Manufacturer: Alfa Aesar™ B2441914

Quantity 25g

 View more versions of this product

Catalog No. AAB2441914

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 106-58-1
    Molecular Formula C6H14N2
    Formula Weight 114.19
    MDL Number MFCD00006156
    UN Number UN2733
    Beilstein 103022
    Chemical Name or Material 1,4-Dimethylpiperazine

    Specifications

    Formula Weight 114.19
    Molecular Formula C6H14N2
    UN Number UN2733
    Health Hazard 2 GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapour.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Flash Point 21°C (69°F)
    Odor Amine-like
    Quantity 25g
    pH 10
    TSCA TSCA
    MDL Number MFCD00006156
    CAS 106-58-1
    Beilstein 103022
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
    RTECS Number TL5945000
    Boiling Point 131° to 132°C
    Melting Point −1°C
    Chemical Name or Material 1,4-Dimethylpiperazine
    Assay 98%

    GHS H Statement
    H225-H314-H318
    Highly flammable liquid and vapour.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit