Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dioxa-8-azaspiro[4.5]decane, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
177-11-7
C7H13NO2
143.19
MFCD00005976
KPKNTUUIEVXMOH-UHFFFAOYSA-N
1,4-dioxa-8-azaspiro 4.5 decane,4-piperidone ethylene ketal,4-piperidinone ethyl ketal,4-piperidone-ethylene ketal,4-piperidone ethylene acetal,1,4-dioxa-8-azaspiro 4,5 decane,4-piperidoneethyleneketal,piperidone-4-ethyleneketal,1,4-dioxa-8-aza-spiro 4.5 decane
67435
1,4-dioxa-8-azaspiro[4.5]decane
C1CNCCC12OCCO2
177-11-7
C7H13NO2
143.19
MFCD00005976
KPKNTUUIEVXMOH-UHFFFAOYSA-N
1,4-dioxa-8-azaspiro 4.5 decane,4-piperidone ethylene ketal,4-piperidinone ethyl ketal,4-piperidone-ethylene ketal,4-piperidone ethylene acetal,1,4-dioxa-8-azaspiro 4,5 decane,4-piperidoneethyleneketal,piperidone-4-ethyleneketal,1,4-dioxa-8-aza-spiro 4.5 decane
67435
1,4-dioxa-8-azaspiro[4.5]decane
C1CNCCC12OCCO2
Specific Gravity 1.11
Flash Point 81°C
Packaging Glass bottle
Infrared Spectrum Authentic
Refractive Index 1.4809 to 1.4829
Boiling Point 108.0°C to 110.0°C (26.0 mmHg)
Color Colorless to Yellow
Quantity 50g
Density 1.1100g/mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 143.19
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1,4-Dioxa-8-azaspiro[4.5]decane, 98%
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 205-868-6

TSCA : TSCA