Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dioxane, 99.5%, Extra Dry over Molecular Sieve, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
1L
2.5L
Packaging:
AcroSeal™ Glass Bottle
Acroseal Glass Bottle
123-91-1
C4H8O2
88.106
MFCD00006571
RYHBNJHYFVUHQT-UHFFFAOYSA-N
dioxane,p-dioxane,1,4-diethylene dioxide,diethylene ether,dioxan,1,4-dioxacyclohexane,diethylene dioxide,dioxanne,di ethylene oxide,tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
C1COCCO1
123-91-1
C4H8O2
88.106
MFCD00006571
RYHBNJHYFVUHQT-UHFFFAOYSA-N
dioxane,p-dioxane,1,4-diethylene dioxide,diethylene ether,dioxan,1,4-dioxacyclohexane,diethylene dioxide,dioxanne,di ethylene oxide,tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
C1COCCO1
CAS Min % 99.4
CAS Max % 100.0
Acetic Acid 0.002 % max.
Assay Percent Range 99.5%
UN Number 1165
Beilstein 19, 3
Fieser 01,333
Merck Index 15, 3328
Formula Weight 88.11
Physical Form Liquid
Percent Purity 99.5%
Grade Extra Dry over Molecular Sieve
Density 1.0340g/mL
Infrared Spectrum Authentic
Packaging AcroSeal™ Glass Bottle
Peroxides 0.005 % max. (as H2O2)
Refractive Index 1.4205 to 1.4220
Solubility Solubility in water: soluble. Other solubilities: soluble in ethanol, acetone, benzene, toluene,, ether and other common organic solvents
Specific Gravity 1.034
Stabilizer 2 to 5ppm BHT
Water 0.005% Max.
Flash Point 12°C
Viscosity 1.32 mPa.s (20°C)
Boiling Point 101.0°C
Color Yellow to Brown
Melting Point 12.0°C
Quantity 2.5L
Chemical Name or Material 1, 4-Dioxane, Over Molecular Sieves
Show More Show Less

GHS H Statement
Causes serious eye irritation. Suspected of causing cancer. May cause respiratory irritation. Highly flammable liquid and vapor. May form explosive peroxides. Repeated exposure may cause skin dryness or cracking.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Keep container tightly closed. Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger