Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
1,4-Dioxane, ≥99.5% (GC), MilliporeSigma™ Supelco™
1,4-Dioxane is an organic compound and an environmental contaminant, generally found in industrial waste streams and by-products. It is commonly used as an industrial solvent, in cosmetic products and household products such as lotions, soap, shampoos, bath foams.
Supplier: MilliporeSigma Supelco 7688710MLF
Description
Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal CareSpecifications
10 mL | |
Analytical Standard | |
MFCD00006571 | |
88.11 | |
Analytical Standard | |
27 mmHg (20°C) | |
1.034 g/mL (at 25°C) | |
100°C to 102°C | |
Neat |
123-91-1 | |
C4H8O2 | |
Diethylene oxide; Dioxane | |
≥99.5% (GC) | |
356°F | |
3 (vs. air) | |
10°C to 12°C | |
12°C |
Safety and Handling
ShelfLife : Limited shelf life, expiry date on the label
RTECSNumber : JG8225000
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction