Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dioxane, B&J Brand™, for HPLC, GC and spectrophotometry, >99.9%, Honeywell Burdick & Jackson

Catalog Number p-7111738
Click to view available options
Quantity:
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
Poly-coated Bottle
123-91-1
C4H8O2
88.106
RYHBNJHYFVUHQT-UHFFFAOYSA-N
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
C1COCCO1
123-91-1
C4H8O2
88.106
RYHBNJHYFVUHQT-UHFFFAOYSA-N
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
C1COCCO1
Absorbance 0.3 AU max. at 250nm (UV), 0.02 AU max. at 300nm (UV), 0.005 AU max. at 350nm (UV), 0.005 AU max. at 400nm (UV)
CAS Max % 100%
UN Number UN1165
Beilstein 102551
Solubility Information Miscible in Water at 20°C
Formula Weight 88.11
Percent Purity >99.9%
Grade HPLC
Density 1.0280 g/cm3
Boiling Point 101.32°C
Quantity 6 x 1 L
Flash Point 12°C
Packaging Glass Bottle
Water 0.05% max.
Chemical Name or Material 1,4-Dioxane, B&J Brand™, for HPLC, GC and spectrophotometry, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Dioxane
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit