Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Dioxane (Histological), Fisher Chemical

Catalog Number d56s4
Click to view available options
Quantity:
4 L
Packaging:
Aluminum Can
123-91-1
C4H8O2
88.106
MFCD00006571
RYHBNJHYFVUHQT-UHFFFAOYSA-N
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
C1COCCO1
123-91-1
C4H8O2
88.106
MFCD00006571
RYHBNJHYFVUHQT-UHFFFAOYSA-N
dioxane, p-dioxane, 1,4-diethylene dioxide, diethylene ether, dioxan, 1,4-dioxacyclohexane, diethylene dioxide, dioxanne, di ethylene oxide, tetrahydro-p-dioxin
31275
CHEBI:47032
1,4-dioxane
C1COCCO1
pH 6 to 8
Melting Point 12°C
Color Colorless
Density 1.034g/cm³
Boiling Point 101°C
Quantity 4 L
Identification Pass Test
Packaging Aluminum Can
Physical Form Liquid
Vapor Pressure 41mbar at 20°C
Viscosity 1.32 mPa/s at 20°C
Grade Histological
Chemical Name or Material 1, 4-Dioxane
Show More Show Less
Hazard Category SIGNAL WORD: DANGER!
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May form explosive peroxides. Possible cancer hazard. May cause cancer based on animal data. Irritating to eyes, respiratory system and skin. Inhalation may cause central nervous system effects. Repeated exposure may cause skin dryness or cracking. Hygroscopic.
NFPA
Health: 2
Flammability: 3
Instability: 1
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit