1,4-Dioxane, HPLC Grade, 99% min, Alfa Aesar™

Manufacturer: Alfa Aesar 22913K2

Quantity 1L

 View more versions of this product

Catalog No. AA22913K2

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 123-91-1
    Physical Form Liquid
    Assay Percent Range 99% min.
    Molecular Formula C4H8O2
    Formula Weight 88.11
    MDL Number MFCD00006571
    Chemical Name or Material 1, 4-Dioxane, Filtered through 0.2μ filters.
    Grade HPLC Grade

    Specifications

    Physical Form Liquid
    Color Clear colorless
    Formula Weight 88.11
    Quantity 1L
    UN Number 1165
    DOT Information DOT Class: 3, Packaging Group: II
    EINECS Number 204-661-8
    Ignition Point 375°C (707°F)
    Odor Ether like
    Vapor Pressure 41 hPa (31mm Hg) at 20°C
    Appearance Colorless liquid
    Assay Percent Range 99% min.
    Molecular Formula C4H8O2
    MDL Number MFCD00006571
    CAS 123-91-1
    Concentration or Composition (by Analyte or Components) 1, 4-Dioxane
    Health Hazard 1 H225-H351-H319-H335: Highly flammable liquid and vapour. Suspected of causing cancer. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material 1, 4-Dioxane, Filtered through 0.2μ filters.
    Grade HPLC Grade

    H225-H351-H319-H335: Highly flammable liquid and vapour. Suspected of causing cancer. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit