Alfa Aesar™ 1,4-Dioxene, 98%

Manufacturer: Alfa Aesar™ H3359714

Quantity 25g

 View more versions of this product

Catalog No. AAH3359714

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 543-75-9
    Molecular Formula C4H6O2
    Formula Weight 86.09
    MDL Number MFCD00082537
    UN Number UN1993
    Synonym 2,3-Dihydro-1,4-dioxin
    Chemical Name or Material 1,4-Dioxene

    Specifications

    Formula Weight 86.09
    Molecular Formula C4H6O2
    UN Number UN1993
    Synonym 2,3-Dihydro-1,4-dioxin
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Boiling Point 94° to 96°C
    MDL Number MFCD00082537
    CAS 543-75-9
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Flash Point 1°C (34°F)
    Quantity 25g
    Chemical Name or Material 1,4-Dioxene
    Assay 98%

    Description

    Applications
    It is ideally suited for use in closed-loop applications such as vented reactors for the manufacture of active pharmaceutical ingredients. Also find serviceable application in the synthesis of this useful heterocyclic intermediate.

    Notes
    Store in cool location. With adequate ventilation. Store under nitrogen. Store away from oxidizing agent.

    GHS H Statement
    H225-H315-H319-H335
    Highly flammable liquid and vapor.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit