Learn More
1,4-Dithio-DL-threitol, 98%, Thermo Scientific Chemicals
Description
Useful for stabilizing sulfhydryl-containing enzymes1,4-Dithio-DL-threitol is used as a protective agent in preventing oxidation of thiol groups and reagent for the reduction of disulfide groups in proteins. It is employed to investigate a-ketoglutarate dehydrogenase function, flavin cofactors in proteins and protein moieties involved in peroxide reduction. It is also useful for stabilizing sulfhydryl-containing enzymes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Useful for stabilizing sulfhydryl-containing enzymes1,4-Dithio-DL-threitol is used as a protective agent in preventing oxidation of thiol groups and reagent for the reduction of disulfide groups in proteins. It is employed to investigate a-ketoglutarate dehydrogenase function, flavin cofactors in proteins and protein moieties involved in peroxide reduction. It is also useful for stabilizing sulfhydryl-containing enzymes.
Solubility
Soluble in water, ethanol, chloroform, diethyl ether, acetone, ether and ethyl acetate.
Notes
Air sensitive and hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents, reducing agents and alkalies.
Specifications
Melting Point | 40°C to 43°C |
Boiling Point | 125°C to 130°C (12 mmHg) |
Flash Point | >110°C (230°F) |
Odor | Unpleasant |
Quantity | 1 g |
Beilstein | 1719757 |
Sensitivity | Air sensitive |
Merck Index | 14,3376 |
Solubility Information | Soluble in water,ethanol,chloroform,diethyl ether,acetone,ether and ethyl acetate. |
Formula Weight | 154.25 |
Show More |
Safety and Handling
Hazard Category | H302-H315-H319-H335 |
Hazard Statement | GHS H Statement H302-H315-H319-H335 Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
EINECSNumber | 222-468-7 |
RTECSNumber | EK1610000 |
TSCA | Yes |
Recommended Storage | Keep cold; Store under Argon |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.