Alfa Aesar™ 1,4-Dithioerythritol, 99%

Manufacturer: Alfa Aesar™ A1013806

Quantity 5g

 View more versions of this product

Catalog No. AAA1013806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6892-68-8
    Molecular Formula C4H10O2S2
    Formula Weight 154.25
    MDL Number MFCD00063750
    UN Number UN3335
    Beilstein 1719756
    Synonym Cleland's Reagent DTE; erythro-1,4-Dimercapto-2,3-butanediol
    Chemical Name or Material 1,4-Dithioerythritol

    Specifications

    Formula Weight 154.25
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    MDL Number MFCD00063750
    CAS 6892-68-8
    UN Number UN3335
    RTECS Number KF2410000
    TSCA TSCA
    Molecular Formula C4H10O2S2
    Beilstein 1719756
    Health Hazard 1 Warning
    Recommended Storage 2°C to 8°C
    Synonym Cleland's Reagent DTE; erythro-1,4-Dimercapto-2,3-butanediol
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 9; Proper Shipping Name: AVIATION REGULATED SOLID, N.O.S.
    Melting Point 82° to 84°C
    Odor Stench
    Quantity 5g
    Chemical Name or Material 1,4-Dithioerythritol
    Assay 99%

    Description

    Applications
    Reagent for maintaining -SH groups in the reduced state, quantitatively reduces disulfides.

    Solubility
    Soluble in water (15.4 mg/ml), and methanol.

    Notes
    Air Sensitive, store away form air. Keep the container tightly sealed. Store away from reducing agent. Store under dry inert gas.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    2°C to 8°C

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit